siRNA Details
Gene Symbol: Dmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022849.3 | NP_074040.2 | 3845 | |||
XM_008759965.1 | XP_008758187.1 | 19, 20 | 2612 | |||
GenBank mRNA | AF540887.1 | - | 3845 | |||
AY548057.1 | - | 3768 | ||||
U32681.1 | - | 3538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
