siRNA Details
Gene Symbol: Fgf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130751.1 | NP_570107.1 | 3 | 334 | ||
GenBank mRNA | AB078902.1 | - | 3 | 334 |

More Information
Other Species Targeted

Gene Ontology Categories:
