siRNA Details
Gene Symbol: Cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130755.1 | NP_570111.1 | 10 | 1131 | ||
GenBank mRNA | AF461496.1 | - | 10 | 1131 | ||
DQ403126.1 | - | 4 | 394 |
Gene Symbol: RGD1565753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765343.1 | XP_008763565.1 | 4 | 1331 |
