siRNA Details
Gene Symbol: Rasgrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130824.2 | NP_570837.2 | 10, 11 | 1420 | ||
GenBank mRNA | AF465263.1 | - | 10, 11 | 1420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
