siRNA Details
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130828.2 | NP_570841.2 | 2, 3 | 447 | ||
GenBank mRNA | AB091379.1 | - | 2, 3 | 386 | ||
AF309631.1 | - | 1, 2 | 125 | |||
AF400653.1 | - | 2, 3 | 167 | |||
AY039651.1 | - | 2, 3 | 447 | |||
BC098649.1 | - | 2, 3 | 429 | 7127895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
