siRNA Details
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131907.2 | NP_571982.2 | 23, 24 | 2598 | ||
GenBank mRNA | BC086994.1 | - | 23, 24 | 2598 | 7106233 | |
M93017.1 | - | 23, 24 | 2309 | |||
M93018.1 | - | 1725 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001253834, NM_175025, NM_001253831, AK161432, AK133703, and others.
Gene Ontology Categories:
