siRNA Details
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270807.1 | NP_001257736.1 | 6 | 1439 | ||
NM_001270808.1 | NP_001257737.1 | 6 | 1488 | |||
NM_001270809.1 | NP_001257738.1 | 6 | 1389 | |||
NM_001270810.1 | NP_001257739.1 | 6 | 2075 | |||
NM_001270811.1 | NP_001257740.1 | 6 | 2425 | |||
NM_133386.3 | NP_596877.2 | 5 | 2679 | |||
XM_006247787.2 | XP_006247849.1 | 6 | 1759 | |||
XM_006247788.2 | XP_006247850.1 | 6 | 1270 | |||
GenBank mRNA | AB049571.1 | - | 6 | 1487 | ||
AB049572.1 | - | 6 | 1439 | |||
AB049573.1 | - | 6 | 1386 | |||
AB049574.1 | - | 6 | 2045 | |||
AB049575.1 | - | 5 | 2192 | |||
BC081738.1 | - | 5 | 2644 | 7132896 | ||
DQ486894.1 | - | 6 | 1227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
