siRNA Details
Gene Symbol: Efna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574979.5 | XP_574979.4 | 2 | 175 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010108, BC107002, BC125003.
Gene Ontology Categories:
