siRNA Details
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133396.2 | NP_596887.1 | 6 | 906 | ||
XM_006238628.2 | XP_006238690.1 | 6 | 1137 | |||
GenBank mRNA | AB049402.1 | - | 6 | 907 | ||
BC081737.1 | - | 7 | 1157 | |||
BC128727.1 | - | 6 | 912 | 7111312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
