siRNA Details
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133404.1 | NP_596895.1 | 16 | 2394 | ||
GenBank mRNA | AB064323.1 | - | 16 | 2394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
