siRNA Details
Gene Symbol: Rap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133410.1 | NP_596901.1 | 1 | 69 | ||
GenBank mRNA | AF386786.1 | - | 1 | 69 |

More Information
Other Species Targeted

Gene Ontology Categories:
