siRNA Details
Gene Symbol: Syf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133417.1 | NP_596908.1 | 6 | 559 | ||
GenBank mRNA | AF366369.1 | - | 6 | 559 | ||
BC081735.1 | - | 5 | 1054 | |||
BC097289.1 | - | 6 | 567 | 7445525 | ||
FQ234066.1 | - | 4 | 386 |

More Information
Other Species Targeted

Gene Ontology Categories:
