siRNA Details
Gene Symbol: Ppp1r14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133425.3 | NP_596916.1 | 2 | 932 | ||
GenBank mRNA | AF407168.3 | - | 2 | 932 | ||
BC086978.1 | - | 2 | 405 | 7191557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
