siRNA Details
Gene Symbol: Hrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133428.2 | NP_596919.1 | 6 | 706 | ||
GenBank mRNA | AB055895.1 | - | 6 | 706 | ||
BC089779.1 | - | 6 | 695 | 7371893 | ||
FQ210661.1 | - | 6 | 706 | |||
FQ219046.1 | - | 4 | 310 |
