siRNA Details
Gene Symbol: Svs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133516.2 | NP_598200.1 | 2 | 204 | ||
GenBank mRNA | X01115.1 | - | 2 | 204 |
Gene Symbol: LOC103694892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773873.1 | XP_008772095.1 | 2 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
