siRNA Details
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133531.2 | NP_598215.1 | 16 | 1929 | ||
GenBank mRNA | AF399873.1 | - | 16 | 1929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
