siRNA Details
Gene Symbol: Mrpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133539.1 | NP_598223.1 | 3 | 314 | ||
GenBank mRNA | BC058447.1 | - | 3 | 356 | 6919782 | |
FQ229319.1 | - | 3 | 369 | |||
U53512.1 | - | 3 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
