siRNA Details
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133544.1 | NP_598228.1 | 2 | 153 | ||
GenBank mRNA | AJ271158.1 | - | 2 | 153 | ||
FQ217274.1 | - | 3 | 160 | |||
FQ217509.1 | - | 3 | 213 | |||
FQ217783.1 | - | 3 | 212 | |||
FQ221212.1 | - | 3 | 211 | |||
FQ223588.1 | - | 3 | 208 | |||
FQ223759.1 | - | 3 | 160 |
Gene Symbol: LOC103693430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768219.1 | XP_008766441.1 | 1 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
