siRNA Details
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133575.1 | NP_598259.1 | 6 | 779 | ||
GenBank mRNA | U49066.1 | - | 6 | 779 |

More Information
Other Species Targeted

Gene Ontology Categories:
