siRNA Details
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133585.3 | NP_598269.3 | 22 | 2332 | ||
XM_006248497.1 | XP_006248559.1 | 24 | 2596 | |||
XM_006248498.1 | XP_006248560.1 | 23 | 2542 | |||
XM_006248499.1 | XP_006248561.1 | 23 | 2485 | |||
GenBank mRNA | AY333988.1 | - | 22 | 2241 | ||
AY510274.1 | - | 22 | 2387 | |||
AY683458.1 | - | 2 | 148 | |||
BC111071.1 | - | 22 | 2332 | 7324017 | ||
U93197.1 | - | 6 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
