siRNA Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191945.1 | NP_001178874.1 | 9 | 1750 | ||
XM_008758887.1 | XP_008757109.1 | 12 | 3303 | |||
GenBank mRNA | D45413.1 | - | 1 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
