siRNA Details
Gene Symbol: Gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133595.2 | NP_598279.1 | 2 | 162 | ||
GenBank mRNA | BC086944.1 | - | 2 | 143 | 7325198 | |
U53710.1 | - | 2 | 162 | |||
U85512.1 | - | 2 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
