siRNA Details
Gene Symbol: Kcne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133603.2 | NP_598287.1 | 2 | 248 | ||
XM_006248023.2 | XP_006248085.1 | 2 | 674 | |||
XM_008768554.1 | XP_008766776.1 | 2 | 484 | |||
GenBank mRNA | AF071003.1 | - | 2 | 89 | ||
BC089778.1 | - | 2 | 248 | 7301610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
