siRNA Details
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276429.1 | NP_001263358.1 | 4 | 227 | ||
NM_001276430.1 | NP_001263359.1 | 3 | 212 | |||
NM_133604.1 | NP_598288.1 | 4 | 298 | |||
GenBank mRNA | CB775939.1 | - | 4 | 277 | ||
FM052814.1 | - | 4 | 136 | |||
FM056491.1 | - | 3 | 175 | |||
FQ228524.1 | - | 4 | 298 |

More Information
Other Species Targeted

Gene Ontology Categories:
