siRNA Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239786.1 | XP_006239848.1 | 11 | 1438 | ||
XM_006239788.1 | XP_006239850.1 | 11 | 1094 | |||
XM_006239789.2 | XP_006239851.1 | 11 | 1369 | |||
GenBank mRNA | BC060545.1 | - | 11 | 1021 | 6918791 | |
D16479.1 | - | 10 | 985 | |||
FM038662.1 | - | 4 | 505 | |||
FM105991.1 | - | 2 | 165 |

More Information
Other Species Targeted

Gene Ontology Categories:
