siRNA Details
Gene Symbol: Gpha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133619.2 | NP_598303.1 | 3 | 323 | ||
XM_008760069.1 | XP_008758291.1 | 3 | 2359 | |||
XM_008760070.1 | XP_008758292.1 | 3 | 2200 | |||
GenBank mRNA | AF260741.1 | - | 3 | 323 |
