siRNA Details
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244480.1 | XP_006244542.1 | 8 | 1069 | ||
GenBank mRNA | AF035952.2 | - | 8 | 1062 | ||
AY035403.1 | - | 8 | 941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
