siRNA Details
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244480.1 | XP_006244542.1 | 1, 2 | 199 | ||
GenBank mRNA | AF035952.2 | - | 1, 2 | 192 | ||
AY035403.1 | - | 1, 2 | 71 |

More Information
Other Species Targeted

Gene Ontology Categories:
