siRNA Details
Gene Symbol: Cox8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134345.1 | NP_599172.1 | 2 | 365 | ||
GenBank mRNA | BC126066.1 | - | 2 | 332 | 7385046 | |
L48209.1 | - | 2 | 283 | |||
X06146.1 | - | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
