siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134346.3 | NP_599173.2 | 5, 6 | 460 | ||
XM_008765377.1 | XP_008763599.1 | 6, 7 | 1020 | |||
GenBank mRNA | AY607847.1 | - | 5, 6 | 460 | ||
BC081731.1 | - | 5, 6 | 605 | 7192310 | ||
FQ229455.1 | - | 5, 6 | 634 | |||
U07795.1 | - | 5, 6 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
