siRNA Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 4 | 319 | ||
GenBank mRNA | BC063150.1 | - | 4 | 317 | 6922267 | |
FQ209427.1 | - | 4 | 340 | |||
FQ209464.1 | - | 4 | 332 | |||
FQ209465.1 | - | 4 | 339 | |||
FQ211049.1 | - | 4 | 339 | |||
FQ212841.1 | - | 4 | 340 | |||
FQ218096.1 | - | 4 | 322 | |||
FQ218830.1 | - | 4 | 347 | |||
FQ219464.1 | - | 4 | 345 | |||
FQ219581.1 | - | 4 | 341 | |||
FQ219850.1 | - | 4 | 331 | |||
J03752.1 | - | 4 | 303 |

More Information
Other Species Targeted

Gene Ontology Categories:
