siRNA Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134351.1 | NP_599178.1 | 4 | 444 | ||
GenBank mRNA | BC062394.1 | - | 477 | 5622019 | ||
J05571.1 | - | 4 | 444 |

More Information
Other Species Targeted

Gene Ontology Categories:
