siRNA Details
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 7 | 1160 | ||
GenBank mRNA | AJ298278.1 | - | 7 | 1160 | ||
BC083176.1 | - | 7 | 1404 | 7097075 | ||
FQ225555.1 | - | 4 | 448 | |||
FQ225622.1 | - | 1 | 56 | |||
FQ226477.1 | - | 2 | 149 | |||
FQ226807.1 | - | 1 | 76 | |||
FQ227215.1 | - | 1 | 98 | |||
FQ227400.1 | - | 2 | 120 | |||
FQ227801.1 | - | 1 | 47 | |||
FQ228174.1 | - | 1 | 61 | |||
FQ230227.1 | - | 2 | 184 | |||
FQ230279.1 | - | 1 | 35 | |||
FQ231126.1 | - | 4 | 448 | |||
FQ231168.1 | - | 4 | 445 | |||
FQ231494.1 | - | 2 | 179 | |||
FQ231724.1 | - | 1 | 72 | |||
FQ232521.1 | - | 1 | 61 | |||
FQ233029.1 | - | 1 | 58 | |||
FQ233189.1 | - | 1 | 73 | |||
FQ233364.1 | - | 4 | 448 | |||
FQ233754.1 | - | 3 | 289 | |||
FQ234227.1 | - | 1 | 59 | |||
FQ234731.1 | - | 4 | 377 | |||
FQ235211.1 | - | 2 | 103 | |||
FQ235339.1 | - | 4 | 437 |

More Information
Other Species Targeted

Gene Ontology Categories:
