siRNA Details
Gene Symbol: Cyp3a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147206.2 | NP_671739.2 | 11 | 1188 | ||
GenBank mRNA | BC086985.1 | - | 11 | 1188 | 7125835 | |
U60085.2 | - | 11 | 1164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
