siRNA Details
Gene Symbol: Ptprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134356.1 | NP_599183.1 | 17, 18 | 2819 | ||
GenBank mRNA | AY177703.1 | - | 17, 18 | 2819 | ||
AY177704.1 | - | 16, 17 | 2732 | |||
AY177705.1 | - | 17, 18 | 2819 | |||
U57501.1 | - | 14 |
