siRNA Details
Gene Symbol: Snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134358.1 | NP_599185.1 | 7 | 1044 | ||
GenBank mRNA | BC083694.1 | - | 7 | 1441 | ||
BC158686.1 | - | 7 | 1044 | 8374870 | ||
FQ214448.1 | - | 7 | 1020 | |||
FQ220404.1 | - | 7 | 997 | |||
FQ220639.1 | - | 7 | 1060 | |||
FQ220740.1 | - | 7 | 1045 | |||
FQ220961.1 | - | 7 | 1045 | |||
FQ229287.1 | - | 7 | 1047 | |||
FQ229512.1 | - | 7 | 1060 | |||
FQ230125.1 | - | 7 | 997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
