siRNA Details
Gene Symbol: Xcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134361.1 | NP_599188.1 | 3 | 290 | ||
GenBank mRNA | U23377.1 | - | 3 | 290 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Xcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134361.1 | NP_599188.1 | 3 | 290 | ||
GenBank mRNA | U23377.1 | - | 3 | 290 |