siRNA Details
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134365.2 | NP_599192.1 | 4 | 399 | ||
GenBank mRNA | BC063808.1 | - | 4 | 340 | 6920021 | |
FQ209675.1 | - | 4 | 298 | |||
FQ212544.1 | - | 4 | 335 | |||
FQ214336.1 | - | 4 | 327 | |||
FQ214371.1 | - | 4 | 335 | |||
FQ214449.1 | - | 4 | 298 | |||
FQ214672.1 | - | 4 | 334 | |||
FQ214784.1 | - | 4 | 333 | |||
FQ215244.1 | - | 4 | 298 | |||
FQ215436.1 | - | 4 | 298 | |||
FQ215480.1 | - | 4 | 298 | |||
FQ215788.1 | - | 4 | 334 | |||
FQ215823.1 | - | 4 | 351 | |||
FQ216062.1 | - | 4 | 300 | |||
FQ216247.1 | - | 4 | 284 | |||
FQ216597.1 | - | 4 | 290 | |||
FQ216773.1 | - | 4 | 298 | |||
FQ216829.1 | - | 4 | 298 | |||
FQ216922.1 | - | 4 | 298 | |||
FQ218701.1 | - | 4 | 334 | |||
FQ219899.1 | - | 4 | 327 | |||
FQ223652.1 | - | 4 | 298 | |||
FQ223683.1 | - | 4 | 324 | |||
FQ229976.1 | - | 4 | 335 | |||
FQ232356.1 | - | 4 | 335 | |||
M35052.1 | - | 4 | 318 |
Gene Symbol: LOC100911417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749372.2 | XP_003749420.1 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
