siRNA Details
Gene Symbol: Rnf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134374.2 | NP_599201.1 | 2, 3 | 562 | ||
GenBank mRNA | AB070897.1 | - | 2, 3 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
