siRNA Details
Gene Symbol: Spata19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134384.1 | NP_599211.1 | 1, 2 | 110 | ||
GenBank mRNA | AB057408.1 | - | 1, 2 | 110 | ||
BC081729.1 | - | 1, 2 | 127 | 7132055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
