siRNA Details
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277260.1 | NP_001264189.1 | 11 | 590 | ||
NM_001277261.2 | NP_001264190.1 | 10 | 557 | |||
NM_001277262.2 | NP_001264191.1 | 10 | 560 | |||
NM_134388.2 | NP_599215.2 | 11 | 593 | |||
GenBank mRNA | AF399874.1 | - | 10 | 541 | ||
AY334079.1 | - | 10 | 537 | |||
AY334080.1 | - | 10 | 534 | |||
AY334081.1 | - | 9 | 504 | |||
AY334082.1 | - | 9 | 501 | |||
FQ215948.1 | - | 9 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
