siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 5 | 1146 | ||
NM_001270590.1 | NP_001257519.1 | 5 | 831 | |||
NM_001270591.1 | NP_001257520.1 | 6 | 609 | |||
NM_001270592.1 | NP_001257521.1 | 6 | 606 | |||
NM_001270593.1 | NP_001257522.1 | 5 | 543 | |||
NM_001270594.1 | NP_001257523.1 | 5 | 540 | |||
NM_134390.3 | NP_599217.1 | 5 | 1149 | |||
XM_006236455.2 | XP_006236517.1 | 5 | 532 | |||
GenBank mRNA | AF370882.1 | - | 4 | 502 | ||
BC059116.1 | - | 5 | 648 | 6920949 | ||
FM070640.1 | - | 5 | 530 | |||
FM086181.1 | - | 6 | 593 | |||
FN804490.1 | - | 6 | 592 | |||
FQ209634.1 | - | 5 | 537 | |||
FQ212987.1 | - | 5 | 538 | |||
FQ219149.1 | - | 6 | 593 | |||
FQ219532.1 | - | 6 | 593 | |||
FQ219576.1 | - | 5 | 537 | |||
FQ220000.1 | - | 5 | 723 | |||
FQ233370.1 | - | 6 | 603 | |||
FQ235102.1 | - | 5 | 555 |
