siRNA Details
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134400.2 | NP_599227.2 | 10, 11 | 766 | ||
XM_006235512.2 | XP_006235574.1 | 11, 12 | 860 | |||
XM_006235513.2 | XP_006235575.1 | 11, 12 | 809 | |||
GenBank mRNA | AF348701.1 | - | 10, 11 | 735 | ||
BC100640.1 | - | 10, 11 | 766 | 7929876 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133763, AK185487, AB045974, AK168357, AK145239, and others.
Gene Ontology Categories:
