siRNA Details
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF063102.1 | - | 5 | 574 | ||
AF081148.1 | - | 6 | 1020 | |||
AF081149.1 | - | 6 | 1020 | |||
AF081150.1 | - | 6 | 1020 | |||
AF081151.1 | - | 6 | 1020 | |||
AF081152.1 | - | 6 | 1020 | |||
AF081153.1 | - | 6 | 1020 | |||
CB579130.1 | - | 4 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
