siRNA Details
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134460.1 | NP_604455.1 | 1368 | |||
GenBank mRNA | BC166845.1 | - | 1368 | 9029827 |

More Information
Other Species Targeted

Gene Ontology Categories:
