siRNA Details
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134460.1 | NP_604455.1 | 6, 7 | 763 | ||
GenBank mRNA | BC166845.1 | - | 6, 7 | 763 | 9029827 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009677, BC052703, AK145821, BC054535, AK034198, and others.
Gene Ontology Categories:
