siRNA Details
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134462.1 | NP_604457.1 | 17 | 1653 | ||
XM_008772595.1 | XP_008770817.1 | 11 | 3633 | |||
XM_008772596.1 | XP_008770818.1 | 11 | 1595 | |||
GenBank mRNA | AF484685.1 | - | 17 | 1653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
