siRNA Details
Gene Symbol: Calcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138513.1 | NP_612522.1 | 2 | 156 | ||
XM_006230075.2 | XP_006230137.1 | 4 | 630 | |||
XM_008759674.1 | XP_008757896.1 | 5 | 485 | |||
GenBank mRNA | M11596.1 | - | 2 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
