siRNA Details
Gene Symbol: LOC171573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138537.2 | NP_612546.1 | 2 | 134 | ||
GenBank mRNA | AF198442.1 | - | 2 | 116 | ||
CB695439.1 | - | 2 | 155 | |||
FQ222501.1 | - | 2 | 154 | |||
FQ228610.1 | - | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
