siRNA Details
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191951.1 | NP_001178880.1 | 7 | 13427 | ||
XM_008760214.1 | XP_008758436.1 | 6 | 13828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
